Learn More
AdipoGen 7-OH-4-methyl-21H-quinolone
SDP
Catalog No. 502560808
Encompass_Preferred
Chemical. CAS 20513-71-7. Formula C10H9NO2. MW 175.18. Synthetic. 7-Hydroxy-4-methyl-21H-quinolone is a starting material for the synthesis of a potential cytotoxic compound class with activity against human tumor cell lines.
Catalog No. 50-256-0808 Supplier AdipoGen Supplier No. CDXH0010M200
Chemical. CAS 20513-71-7. Formula C10H9NO2. MW 175.18. Synthetic. 7-Hydroxy-4-methyl-21H-quinolone is a starting material for the synthesis of a potential cytotoxic compound class with activity against human tumor cell lines.