Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-oxo-7H-benzimidazo[2,1-a]benzo[de]isoquinoline-3-carboxylic acid | 52029-86-4 | MFCD30685721 | 98.2% | 314.29 | C19H10N2O3 | 10 MG
SDP

Catalog No. 5000297549
Encompass_Preferred

STO-609 is a selective, cell-permeable inhibitor of calmodulin-dependent protein kinase kinase (CaMKK) used as a research tool to probe CaMKK/CaMK and AMPK signaling pathways in biochemical and cell-based studies. The compound is supplied as a solid research reagent with documented purity, solubility, and storage recommendations for laboratory use.

  • Selective inhibitor of CaMKK with activity against CaMKKα and CaMKKβ.
  • Cell-permeable small molecule suitable for cellular assays.
  • Useful probe for AMPK and autophagy pathway studies.
  • Purity 98.2% by HPLC.
  • Molecular weight 314.29; formula C19H10N2O3; CAS 52029-86-4.
  • Available in small research sizes, including 10 mg.
  • Solubility varies by vehicle; soluble in DMSO and PEG-containing formulations.
  • Store sealed at 4°C; in solvent, store at -80°C for long-term stability.

Catalog No. 50-002-97549 Supplier Medchemexpress LLC Supplier No. HY1980510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

STO-609 is a selective, cell-permeable inhibitor of calmodulin-dependent protein kinase kinase (CaMKK) used as a research tool to probe CaMKK/CaMK and AMPK signaling pathways in biochemical and cell-based studies. The compound is supplied as a solid research reagent with documented purity, solubility, and storage recommendations for laboratory use.

  • Selective inhibitor of CaMKK with activity against CaMKKα and CaMKKβ.
  • Cell-permeable small molecule suitable for cellular assays.
  • Useful probe for AMPK and autophagy pathway studies.
  • Purity 98.2% by HPLC.
  • Molecular weight 314.29; formula C19H10N2O3; CAS 52029-86-4.
  • Available in small research sizes, including 10 mg.
  • Solubility varies by vehicle; soluble in DMSO and PEG-containing formulations.
  • Store sealed at 4°C; in solvent, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.