Learn More
Medchemexpress LLC 7-oxodehydroabietic acid | 18684-55-4 | MFCD01699051 | 99.7% | 314.42 g·mol⁻1 | C20H26O3 | 5 MG
SDP
Catalog No. 5000262583
Encompass_Preferred
7-oxodehydroabietic acid (CAS 18684-55-4) is a diterpene resin acid used in natural product research and studies of insect endocrine-disrupting activity. It is supplied as a white to off-white solid with high reported purity for laboratory research applications.
  • High purity: 99.7% as supplied on the COA.
  • White to off-white solid suitable for analytical and preparative work.
  • Molecular formula C20H26O3 and molecular weight 314.42 g·mol⁻1.
  • Available in small milligram pack sizes for research use.
  • Certificate of analysis and safety data sheet available for quality and safety verification.
Catalog No. 50-002-62583 Supplier Medchemexpress LLC Supplier No. HY1336205MG
7-oxodehydroabietic acid (CAS 18684-55-4) is a diterpene resin acid used in natural product research and studies of insect endocrine-disrupting activity. It is supplied as a white to off-white solid with high reported purity for laboratory research applications.
  • High purity: 99.7% as supplied on the COA.
  • White to off-white solid suitable for analytical and preparative work.
  • Molecular formula C20H26O3 and molecular weight 314.42 g·mol⁻1.
  • Available in small milligram pack sizes for research use.
  • Certificate of analysis and safety data sheet available for quality and safety verification.