Learn More
Cliawaived Inc CLIAwaived, Inc. 7 Panel - IDTC II
SDP
Catalog No. NC1859230
Encompass_Preferred
(25 Cups)CLIAwaived, Inc. - INSTANT DRUG TEST CUP/CARD II (IDTC II). This 7-panel, no-leak drug test cup tests for BZO, COC, MET, MTD, THC, OPI (MOR 300) and OXY. Results in 5 minutes.FDA-cleared and CLIA-waived.
This item is not returnable. View return policy
Catalog No. NC1859230 Supplier Cliawaived Inc Supplier No. CLIAIDTC760717
(25 Cups)CLIAwaived, Inc. - INSTANT DRUG TEST CUP/CARD II (IDTC II). This 7-panel, no-leak drug test cup tests for BZO, COC, MET, MTD, THC, OPI (MOR 300) and OXY. Results in 5 minutes.FDA-cleared and CLIA-waived.