Learn More
BTNX Inc 7 Parameter Urinalysis Reagent Strips 100pk
Catalog No. NC3361246
Encompass_Preferred
The Rapid Response Urinalysis Reagent Strips are qualitative and semi-qualitative dip-and-read test for the detection of urine analytes in human urine, to aid in the general evaluation of health and in the diagnosis and monitoring of metabolic or systemic diseases.The 7 Parameter Rapid Response Urinalysis Reagent Strips simultaneously detects Glucose, Ketone, Protein, pH, Blood, Nitrite and Leukocytes in human urine. Compatable with the Rapid Response Urine Analyzer U120.
This item is not returnable. View return policy
Catalog No. NC3361246 Supplier BTNX Inc Supplier No. U7.11S100
The Rapid Response Urinalysis Reagent Strips are qualitative and semi-qualitative dip-and-read test for the detection of urine analytes in human urine, to aid in the general evaluation of health and in the diagnosis and monitoring of metabolic or systemic diseases.The 7 Parameter Rapid Response Urinalysis Reagent Strips simultaneously detects Glucose, Ketone, Protein, pH, Blood, Nitrite and Leukocytes in human urine. Compatable with the Rapid Response Urine Analyzer U120.