Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-pentadecanol | 4104-59-0 | MFCD00046732 | 98.7% | 228.41 g·mol⁻¹ | C15H32O | 10 G
SDP

Catalog No. 5000284573
Encompass_Preferred

7-Pentadecanol (pentadecan-7-ol) is a long-chain fatty alcohol reagent used in lipid research, analytical standards, and synthetic chemistry. It is a white to off-white solid with a hydroxyl functional group at the C7 position and is supplied with analytical data and storage guidance for research use.

  • High purity: 98.7% (GC).
  • Consistent composition: molecular formula C15H32O, MW 228.41 g·mol⁻¹.
  • Solid, white to off-white appearance for straightforward handling and weighing.
  • Stable under recommended storage: room temperature (solid), solvent storage at -20 °C to -80 °C for extended retention.
  • Analytical support provided: 1H NMR consistent with structure; optical rotation reported.

Catalog No. 50-002-84573 Supplier Medchemexpress LLC Supplier No. HYW59483310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Pentadecanol (pentadecan-7-ol) is a long-chain fatty alcohol reagent used in lipid research, analytical standards, and synthetic chemistry. It is a white to off-white solid with a hydroxyl functional group at the C7 position and is supplied with analytical data and storage guidance for research use.

  • High purity: 98.7% (GC).
  • Consistent composition: molecular formula C15H32O, MW 228.41 g·mol⁻¹.
  • Solid, white to off-white appearance for straightforward handling and weighing.
  • Stable under recommended storage: room temperature (solid), solvent storage at -20 °C to -80 °C for extended retention.
  • Analytical support provided: 1H NMR consistent with structure; optical rotation reported.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.