Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-pentadecanol | 4104-59-0 | MFCD00046732 | 98.7% | 228.41 g/mol | C15H32O | 250 MG
SDP

Catalog No. 5000284503
Encompass_Preferred

7-Pentadecanol is a long-chain fatty alcohol used as a biochemical assay reagent and research chemical. It has a terminal hydroxyl group on a 15-carbon hydrocarbon chain (C15H32O) and a molecular weight of 228.41 g/mol. Supplied as a white to off-white solid with reported purity 98.72% (GC). For laboratory research use only; not for human or veterinary use.

  • Reported purity 98.72% (GC) suitable for analytical and assay work.
  • Primary alcohol functional group for lipid and membrane studies.
  • Solid form simplifies handling and weighing for experiments.
  • Storage stability: room temperature (3 years); in solvent -80°C (2 years), -20°C (1 year).
  • Chemical formula C15H32O; molecular weight 228.41 g/mol.

Catalog No. 50-002-84503 Supplier Medchemexpress LLC Supplier No. HYW594833250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Pentadecanol is a long-chain fatty alcohol used as a biochemical assay reagent and research chemical. It has a terminal hydroxyl group on a 15-carbon hydrocarbon chain (C15H32O) and a molecular weight of 228.41 g/mol. Supplied as a white to off-white solid with reported purity 98.72% (GC). For laboratory research use only; not for human or veterinary use.

  • Reported purity 98.72% (GC) suitable for analytical and assay work.
  • Primary alcohol functional group for lipid and membrane studies.
  • Solid form simplifies handling and weighing for experiments.
  • Storage stability: room temperature (3 years); in solvent -80°C (2 years), -20°C (1 year).
  • Chemical formula C15H32O; molecular weight 228.41 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.