Learn More
Cayman Chemical 7-(p-Methoxybenzylamino)-4-nitrobenz-2,1,3-oxadiazole, Purity: ≥98%, CAS: 33984-50-8, 50 mg
SDP
Catalog No. 502919196
Encompass_Preferred
A fluorescent probe for hydrophobic regions of proteins; upon binding to a hydrophobic region of a protein, displays an emission of 530 nm upon excitation at 430 nm
Catalog No. 50-291-9196 Supplier Cayman Chemical Supplier No. 3966850 MG
A fluorescent probe for hydrophobic regions of proteins; upon binding to a hydrophobic region of a protein, displays an emission of 530 nm upon excitation at 430 nm