Learn More
Waters Corp Valve Cartridge Assembly, 7-port, for ACQUITY Column Manager, UPLC System
Catalog No. 502391199 Shop All Waters Corporation Products
Encompass_Preferred
The 7-port Valve Cartridge Assembly serves as a replacement part within the original Left/Right Inlet ACQUITY Column Manager, and is designed for use in UPLC Systems.
  • Functions as an injector/valve pod.
  • Designed as a spare part kit.
  • Includes 1 unit per package.
  • Compatible with UPLC systems.
  • Replaced as required for maintenance.
Catalog No. 50-239-1199 Supplier Waters Corp Supplier No. 700003145
The 7-port Valve Cartridge Assembly serves as a replacement part within the original Left/Right Inlet ACQUITY Column Manager, and is designed for use in UPLC Systems.
  • Functions as an injector/valve pod.
  • Designed as a spare part kit.
  • Includes 1 unit per package.
  • Compatible with UPLC systems.
  • Replaced as required for maintenance.