Learn More
Medchemexpress LLC 7-PPD | 3081-01-4 | 98.28% | 282.42 | 25 G
SDP
Catalog No. 5000384438
Encompass_Preferred
7-PPD (DMPPD) is a rubber antioxidant that scavenges free radicals and inhibits the oxidative degradation of rubber chains. It shows promise for research in tire manufacturing and rubber product processing.
  • Scavenges free radicals.
  • Inhibits oxidative degradation of rubber chains.
  • Potential for use in tire manufacturing.
  • Suitable for rubber product processing research.
Catalog No. 50-003-84438 Supplier Medchemexpress LLC Supplier No. 25GHY176747
7-PPD (DMPPD) is a rubber antioxidant that scavenges free radicals and inhibits the oxidative degradation of rubber chains. It shows promise for research in tire manufacturing and rubber product processing.
  • Scavenges free radicals.
  • Inhibits oxidative degradation of rubber chains.
  • Potential for use in tire manufacturing.
  • Suitable for rubber product processing research.