Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Quinolinol 98.0+%, TCI America™
SDP

Catalog No. Q00841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q00841G Supplier TCI America Supplier No. Q00841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 580-20-1
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00016730
InChI Key QUUFTOGCVYWGNK-UHFFFAOYSA-N
Synonym 7-hydroxyquinoline, quinolin-7-ol, 7-quinolinol, unii-d2g6yuk7zj, 7-chinolinol, ccris 4332, d2g6yuk7zj, xcrppapdrubkrj-uhfffaoysa-n, quinolin-1-ium-7-olate, 7hydroxyquinoline
PubChem CID 11378
IUPAC Name 1H-quinolin-7-one
SMILES C1=CNC2=CC(=O)C=CC2=C1

Specifications

Melting Point 242°C
Quantity 1 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Quinolinol
RTECSNumber VC4160000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.