Learn More
Chemglass Life Sciences Glassblowers Hose Connection, Direct Sealing, 7 Rings, Dimensions (mm): "A" - 10.0, "B" - 22.0, "C" - 27, "D" - 56, "E" Tubing O.D. 25mm
SDP
Supplier:  Chemglass Life Sciences CG30104
Encompass_Preferred
Glassblowers Hose Connection, Direct Sealing, 7 Rings, Dimensions (mm): "A" - 10.0, "B" - 22.0, "C" - 27, "D" - 56, "E" Tubing O.D. 25mm. Connections are tooled to form a flare for sealing directly to apparatus using the tubing as a holder. Serrations are tooled on both ends of an 8" length of tubing. Hose connections are SOLD IN PACKAGES OF 10 SERRATED CONNECTORS (5 pieces tooled on both ends).
Catalog No. CG30104