Learn More
Cayman Chemical 7(S),10(S)-DiHOME, Purity: ≥95%, CAS: 252255-87-1, 10 mg
SDP
Catalog No. 502947634
Encompass_Preferred
An antibacterial hydroxy fatty acid; active against various food-borne pathogenic bacteria (MIC50s = 31.3-250 µg/ml), as well as various plant pathogenic bacteria (MIC90s = 125-500 µg/ml, respectively)
Catalog No. 50-294-7634 Supplier Cayman Chemical Supplier No. 3344710 MG
An antibacterial hydroxy fatty acid; active against various food-borne pathogenic bacteria (MIC50s = 31.3-250 µg/ml), as well as various plant pathogenic bacteria (MIC90s = 125-500 µg/ml, respectively)