Learn More
Cayman Chemical Resolvin D2, 810668-37-2, 25 g
SDP
Supplier:  Cayman Chemical 1000727925
Encompass_Preferred
Molecular Formula C22H32O5, Purity ≥95%, Formula Weight 376.5, Synonyms: 7(S),16(R),17(S)-Resolvin D2, RvD2; Dampens excessive neutrophil trafficking to sites of inflammation; reduces neutrophil infiltration by 70% at doses as low as 10 pg per mouse and significantly reduces leukocyte adherence and emigration at 1 nM.
Catalog No. NC0518452