Learn More
Medchemexpress LLC 7-TFA-ap-7-Deaza-dA | 178420-75-2 | 98.8% | 399.32 | 100 MG
SDP
Catalog No. 5000349691
Encompass_Preferred
7-TFA-ap-7-Deaza-dA is a modified nucleoside and a click chemistry reagent. It is utilized in the synthesis of deoxyribonucleic acid or nucleic acid. Featuring an alkyne group, this compound is capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. It is intended for laboratory use and substance manufacturing, strictly for research purposes.
  • Modified nucleoside
  • Click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Supports DNA and nucleic acid synthesis
  • For laboratory chemicals and substance manufacturing
  • For research use only
Catalog No. 50-003-49691 Supplier Medchemexpress LLC Supplier No. HY138590100MG
7-TFA-ap-7-Deaza-dA is a modified nucleoside and a click chemistry reagent. It is utilized in the synthesis of deoxyribonucleic acid or nucleic acid. Featuring an alkyne group, this compound is capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. It is intended for laboratory use and substance manufacturing, strictly for research purposes.
  • Modified nucleoside
  • Click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Supports DNA and nucleic acid synthesis
  • For laboratory chemicals and substance manufacturing
  • For research use only