Learn More
Medchemexpress LLC 7-TFA-ap-7-Deaza-dA | 178420-75-2 | 98.8% | 399.32 | 50 MG
SDP
Catalog No. 5000347866
Encompass_Preferred
7-TFA-ap-7-Deaza-dA is a modified nucleoside that can be utilized in the synthesis of deoxyribonucleic acid or nucleic acid. It functions as a click chemistry reagent due to its Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Modified nucleoside for nucleic acid synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Related to nucleoside analogs and adenosine
  • Applicable in cancer research and cell cycle/DNA damage studies
Catalog No. 50-003-47866 Supplier Medchemexpress LLC Supplier No. HY13859050MG
7-TFA-ap-7-Deaza-dA is a modified nucleoside that can be utilized in the synthesis of deoxyribonucleic acid or nucleic acid. It functions as a click chemistry reagent due to its Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Modified nucleoside for nucleic acid synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
  • Related to nucleoside analogs and adenosine
  • Applicable in cancer research and cell cycle/DNA damage studies