Learn More
Medchemexpress LLC 7-tfa-ap-7-deaza-da | 178420-75-2 | 98.8% | C16H16F3N5O4 | 10MG
SDP
Catalog No. 5000260173
Encompass_Preferred
7-TFA-ap-7-Deaza-dA is a modified deoxynucleoside bearing a trifluoroacetyl-protected alkyne; designed for incorporation into DNA/oligonucleotides and for conjugation via copper-catalyzed azide-alkyne cycloaddition. Supplied as a solid or as a DMSO solution for use in synthetic and analytical workflows requiring a terminal alkyne handle.
  • Contains an alkyne functional group for CuAAC click chemistry.
  • Suitable for incorporation into deoxyribonucleic acid and oligonucleotide synthesis.
  • Available as a solid and as a 10 mM solution in DMSO.
  • High purity (~98.8%) suitable for research applications.
  • Store solids at 4°C sealed; store solutions at low temperatures to preserve stability.
Catalog No. 50-002-60173 Supplier Medchemexpress LLC Supplier No. HY13859010MG
7-TFA-ap-7-Deaza-dA is a modified deoxynucleoside bearing a trifluoroacetyl-protected alkyne; designed for incorporation into DNA/oligonucleotides and for conjugation via copper-catalyzed azide-alkyne cycloaddition. Supplied as a solid or as a DMSO solution for use in synthetic and analytical workflows requiring a terminal alkyne handle.
  • Contains an alkyne functional group for CuAAC click chemistry.
  • Suitable for incorporation into deoxyribonucleic acid and oligonucleotide synthesis.
  • Available as a solid and as a 10 mM solution in DMSO.
  • High purity (~98.8%) suitable for research applications.
  • Store solids at 4°C sealed; store solutions at low temperatures to preserve stability.