Learn More
Medchemexpress LLC 7-TFA-ap-7-Deaza-dG | 666847-77-4 | 98.0% | 415.32 | 10 MM / 1 ML
SDP
Catalog No. 5000347865
Encompass_Preferred
7-TFA-ap-7-Deaza-dG is a modified nucleoside and a click chemistry reagent. It contains an alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing azide groups.
  • Modified nucleoside
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Used in DNA/RNA synthesis
  • Functions as a nucleoside antimetabolite/analog
Catalog No. 50-003-47865 Supplier Medchemexpress LLC Supplier No. HY13858910MM/1ML
7-TFA-ap-7-Deaza-dG is a modified nucleoside and a click chemistry reagent. It contains an alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAC) with molecules containing azide groups.
  • Modified nucleoside
  • Click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Used in DNA/RNA synthesis
  • Functions as a nucleoside antimetabolite/analog