Learn More
Medchemexpress LLC 7-TFA-ap-7-Deaza-dG | 666847-77-4 | 98.0% | 415.32 g/mol | C16H16F3N5O5 | 5 MG
SDP
Catalog No. 5000221094
Encompass_Preferred
7-TFA-ap-7-Deaza-dG is a modified nucleoside (7-trifluoroacetyl 7-deaza-2'-deoxyguanosine) designed as a clickable building block for DNA and RNA synthesis and modification. It is provided in solid and solution formats and is accompanied by analytical documents (data sheet, COA, H NMR, RP-HPLC, MS, and SDS) to support research use.
  • Contains an alkyne group enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • High reported purity for reproducible experimental results.
  • Available in multiple formats, including small-scale solid and ready-to-use solutions.
  • Comes with analytical documentation to verify identity and quality.
  • Suitable as a building block or analogue in nucleic acid chemistry and click chemistry workflows.
Catalog No. 50-002-21094 Supplier Medchemexpress LLC Supplier No. HY1385895MG
7-TFA-ap-7-Deaza-dG is a modified nucleoside (7-trifluoroacetyl 7-deaza-2'-deoxyguanosine) designed as a clickable building block for DNA and RNA synthesis and modification. It is provided in solid and solution formats and is accompanied by analytical documents (data sheet, COA, H NMR, RP-HPLC, MS, and SDS) to support research use.
  • Contains an alkyne group enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • High reported purity for reproducible experimental results.
  • Available in multiple formats, including small-scale solid and ready-to-use solutions.
  • Comes with analytical documentation to verify identity and quality.
  • Suitable as a building block or analogue in nucleic acid chemistry and click chemistry workflows.