Learn More
Medchemexpress LLC 7-TFA-ap-7-Deaza-dG | 666847-77-4 | MFCD29904591 | 98.0% | C16H16F3N5O5 | 10MG
SDP
Catalog No. 5000259252
Encompass_Preferred
7-TFA-ap-7-Deaza-dG is a modified nucleoside (7-deaza-2'-deoxyguanosine) bearing a trifluoroacetylated propargyl linker. Supplied as a solid, it functions as a building block in oligonucleotide synthesis and as a reagent for click-chemistry labeling, enabling conjugation through its terminal alkyne for bioconjugation and probe attachment.
  • 7-deaza 2'-deoxyguanosine core with trifluoroacetyl modification.
  • Terminal alkyne (propargyl) handle for click chemistry conjugation.
  • High purity suitable for research and synthesis.
  • Solid form for convenient storage and handling.
  • Compatible with nucleotide and oligonucleotide synthesis workflows.
Catalog No. 50-002-59252 Supplier Medchemexpress LLC Supplier No. HY13858910MG
7-TFA-ap-7-Deaza-dG is a modified nucleoside (7-deaza-2'-deoxyguanosine) bearing a trifluoroacetylated propargyl linker. Supplied as a solid, it functions as a building block in oligonucleotide synthesis and as a reagent for click-chemistry labeling, enabling conjugation through its terminal alkyne for bioconjugation and probe attachment.
  • 7-deaza 2'-deoxyguanosine core with trifluoroacetyl modification.
  • Terminal alkyne (propargyl) handle for click chemistry conjugation.
  • High purity suitable for research and synthesis.
  • Solid form for convenient storage and handling.
  • Compatible with nucleotide and oligonucleotide synthesis workflows.