Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-Thieno[3,2-d]pyrimidine

Catalog No. SYX003651G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003651G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003651G Supplier Sigma Aldrich Supplier No. SYX003651G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Butyl chloroformate was used in the synthesis of dextran alkyl carbonates
  • It was also used in the synthesis of s-tert-alkyl-n,n-alkoxycarbonylmethyl-dithiocarbamate, radical polymerization with reversible addition-fragmentation chain transfer agent
  • It was used as derivatization reagent in determination of linsidomine in plasma using high-performance liquid chromatographic separation with tandem mass spectrometric detection

Specifications

CAS 272-68-4
Molecular Formula C6H4N2S
Linear Formula C6H4N2S
MDL Number MFCD11110439
Quantity 100 g
Molecular Weight (g/mol) 136.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.