Learn More
Medchemexpress LLC 7-tridecanol | 927-45-7 | MFCD00021955 | 99.5% | 200.36 g/mol | C13H28O | 250 MG
SDP
Catalog No. 5000284101
Encompass_Preferred
7-Tridecanol (C13H28O; CAS 927-45-7) is a saturated primary fatty alcohol used as a biochemical assay reagent and research chemical. It is biodegradable, has known structural isomers, and is supplied at laboratory-grade purity for analytical and formulation work.
  • Primary fatty alcohol with formula C13H28O
  • High purity (manufacturer lists ~99.47%)
  • Intended for biochemical assays and research use
  • Biodegradable and chemically stable under typical laboratory conditions
  • Suitable as a reference material for analytical and formulation studies
Catalog No. 50-002-84101 Supplier Medchemexpress LLC Supplier No. HYW578187250MG
7-Tridecanol (C13H28O; CAS 927-45-7) is a saturated primary fatty alcohol used as a biochemical assay reagent and research chemical. It is biodegradable, has known structural isomers, and is supplied at laboratory-grade purity for analytical and formulation work.
  • Primary fatty alcohol with formula C13H28O
  • High purity (manufacturer lists ~99.47%)
  • Intended for biochemical assays and research use
  • Biodegradable and chemically stable under typical laboratory conditions
  • Suitable as a reference material for analytical and formulation studies