Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Tridecanone 97.0+%, TCI America™
SDP

Catalog No. D15465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15465G 5 g
D154625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15465G Supplier TCI America Supplier No. D15465G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 462-18-0
Molecular Formula C13H26O
Molecular Weight (g/mol) 198.35
MDL Number MFCD00009543
InChI Key ULIAPOFMBCCSPE-UHFFFAOYSA-N
Synonym 7-tridecanone, dihexyl ketone, enanthone, hexyl ketone, di-n-hexyl ketone, dihexadecyl ketone, unii-l0rdd0t8dg, l0rdd0t8dg, dihexylketone, di-normal-hexyl ketone
PubChem CID 10015
IUPAC Name tridecan-7-one
SMILES CCCCCCC(=O)CCCCCC

Specifications

Color White-Yellow
Boiling Point 264°C
Quantity 5 g
Formula Weight 198.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 7-Tridecanone
EINECSNumber (2)-2475
RTECSNumber YD4577000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.