Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 7mm ID X 9mm OD
SDP
Catalog No. CGQ031006
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 7mm ID X 9mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031006 Supplier Chemglass Life Sciences Supplier No. CGQ031006
Graded Seal, Quartz to Borosilicate, 7mm ID X 9mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.