Learn More
Medchemexpress LLC 7(Z),10(Z),13(Z)-hexadecatrienoic acid | 7561-64-0 | 98.2% | 250.38 | C16H26O2 | 1mg
SDP
Catalog No. 5000268441
Encompass_Preferred
7(Z) 10(Z) 13(Z)-Hexadecatrienoic acid (Roughanic acid) is an -3 polyunsaturated fatty acid (PUFA) that can be found in the leaves of S olusatrum 7(Z) 10(Z) 13(Z)-Hexadecatrienoic acid is an intermediate in the biosynthesis of jasmonic acid [1][2]
Catalog No. 50-002-68441 Supplier Medchemexpress LLC Supplier No. HYW7183831MG
7(Z) 10(Z) 13(Z)-Hexadecatrienoic acid (Roughanic acid) is an -3 polyunsaturated fatty acid (PUFA) that can be found in the leaves of S olusatrum 7(Z) 10(Z) 13(Z)-Hexadecatrienoic acid is an intermediate in the biosynthesis of jasmonic acid [1][2]