Learn More
Cayman Chemical 7(Z),11(Z)-Pentacosadiene, Purity: ≥98%, CAS: 127599-39-7, 10 mg
SDP
Catalog No. 502893685
Encompass_Preferred
A 25-C diene that is found in low abundance on cuticles of mature Drosophila females; depletion of a female-specific elongase (eloF), which leads to an increase in 7(Z),11(Z)-pentacosadiene along with a parallel decrease in 7,11-nonacosadiene, significantly reduces copulation in Drosophila
Catalog No. 50-289-3685 Supplier Cayman Chemical Supplier No. 900053110 MG
A 25-C diene that is found in low abundance on cuticles of mature Drosophila females; depletion of a female-specific elongase (eloF), which leads to an increase in 7(Z),11(Z)-pentacosadiene along with a parallel decrease in 7,11-nonacosadiene, significantly reduces copulation in Drosophila