Learn More
Cayman Chemical 7 Z 11 ZHeptacosadiene
SDP
Catalog No. 502888588
Encompass_Preferred
Female mating pheromone of D. melanogaster; elicits wing vibrations in male D. melanogaster flies at >100 ng
Catalog No. 50-288-8588 Supplier Cayman Chemical Supplier No. 1001256710 MG
Female mating pheromone of D. melanogaster; elicits wing vibrations in male D. melanogaster flies at >100 ng