Learn More
Cayman Chemical 7(Z)-Pentacosene, Purity: ≥98%, CAS: 63623-49-4, 10 mg
SDP
Catalog No. 502893695
Encompass_Preferred
An unsaturated cuticular hydrocarbon that acts as a contact sex pheromone in Drosophila; may stimulate copulation either by itself at high concentrations or when combined with other alkenes; increases in Drosophila females immediately after copulation
Catalog No. 50-289-3695 Supplier Cayman Chemical Supplier No. 900053010 MG
An unsaturated cuticular hydrocarbon that acts as a contact sex pheromone in Drosophila; may stimulate copulation either by itself at high concentrations or when combined with other alkenes; increases in Drosophila females immediately after copulation