Learn More
Cayman Chemical 7 Z-TrIcosen 10mg
SDP
Catalog No. 502893959
Encompass_Preferred
An unsaturated cuticular hydrocarbon that acts as a pheromone in some insects, including Drosophila; abundant only in males of D. melanogaster and D. sechellia; prevents or reduces male courtship behavior and increases female sexual receptivity
Catalog No. 50-289-3959 Supplier Cayman Chemical Supplier No. 900031310 MG
An unsaturated cuticular hydrocarbon that acts as a pheromone in some insects, including Drosophila; abundant only in males of D. melanogaster and D. sechellia; prevents or reduces male courtship behavior and increases female sexual receptivity