Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

70/30 IPA/Water Solution, Solstice

Catalog No. 60090351 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
200 l
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
60-090-351 200 l
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-351 Supplier Solstice Supplier No. CS420ZA200

missing translation for 'validMainframeMsg'

LabReady™ Solvent Blend, ACS Reagent, 70% Isopropyl Alcohol in Water

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-63-0, 7732-18-5
CAS Max % 70.00%, 30.00%
Quantity 200 L
Flash Point 11.7°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid, clear
Purity Grade Notes Other catalog blends
Boiling Point 87.7°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.