Learn More
Burch Bottle & Packaging Inc 70-400 BLACK POLYPROPYLENE RM
Catalog No. NC1778710
Encompass
Stock number 2BP0706MR is a 70/400 Black Polypropylene injection molded closure. This cap features ribbed sides allowing for easy application/removal and a matte top. It has an F217 (foam) and pressure sensitive (PS-22) liner. Printing on the pressure sensitive liner reads "Sealed for Your Protection".
Catalog No. NC1778710 Supplier Burch Bottle & Packaging Inc Supplier No. 2BP0706MR
Stock number 2BP0706MR is a 70/400 Black Polypropylene injection molded closure. This cap features ribbed sides allowing for easy application/removal and a matte top. It has an F217 (foam) and pressure sensitive (PS-22) liner. Printing on the pressure sensitive liner reads "Sealed for Your Protection".