Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 70% Ethanol Prefills
SDP

Catalog No. 21402131
Change view
Click to view available options
Color:
Clear Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-131 Clear Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-402-131 Supplier StatLab Supplier No. ALREA5CB

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

120 ml ethanol prefills vials with with 30 ml fill for removing water from tissue samples

Histological Uses Include:

  • Dehydration: Solution is used to remove water from tissue samples, preserving them for further processing and staining
  • Rehydration: After dehydration, 70% ethanol is used to gradually rehydrate the tissue samples, preventing cell damage and ensuring optimal staining results
  • Staining: 70% ethanol serves as a solvent for specific stains and as a rinse bath to remove excess dye, ensuring precise staining patterns
  • Fixative Component: It can be used as a component in some fixative formulations or for post-fixation washes

Specifications

Physical Form Solution
CAS 67-63-0, 64-17-5
Color Clear Colorless
Quantity 5 ga.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.