Learn More
Lime Media 70% Ethanol Sanitizer with Aloe, Plant Based Grade
Description
- Green, Plant Base Grade Sanitizer
- Formula includes Aloe
- 70% Ethanol
- 500 Viscosity
- Pallet of 1 Gallon Bottles (192)
Specifications
Specifications
| Product Type | Sanitizing Fluid |
| Form | Liquid |
| Composition | 70% Ethanol with Aloe |
| Packaging | Pallet |
| Volume (English) | 1 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.