Learn More
TRIBIOSCIENCE INC 70% Ethanol Solution (Nuclease Free), 1L
SDP
Supplier:  TRIBIOSCIENCE INC TBS5091
Encompass
Tribioscience’s 70% Ethanol Solution is made of 200 proof ethanol and Ultrapure and DEPC treated water. This solution is also great in combination with commercial “mini-prep” kits, magnetic bead DNA extraction as washing buffer. Applications & Features: Excellent for all RNA and DNA purification, precipitation, or manipulation techniques; Works with all commercial RNA and DNA purification kits; Can be used as a washing buffer in magnetic bead DNA purification approaches; Made with DEPC treated water to eliminate nuclease.
Catalog No. NC3883683