Learn More
BD Cell Analysis 3P BD Biosciences 70 µm Integrated Nozzle, Optimized for BD FACSAria III Cell Sorter, Square Cuvette Design, Unit: Each
Catalog No. NC0446302
Encompass
The BD Biosciences 70 Micron Integrated Nozzle is essential for advanced cell sorting applications using the BD FACSAria III. This precision component features a 70 µm orifice optimized for consistent performance in research settings. Its unique design ensures precise positioning, enhancing sorting accuracy and reliability. The product is ideal for users seeking high-performance solutions in cell analysis and sorting, although it is now discontinued.
Catalog No. NC0446302 Supplier BD Cell Analysis 3P Supplier No. 647339
The BD Biosciences 70 Micron Integrated Nozzle is essential for advanced cell sorting applications using the BD FACSAria III. This precision component features a 70 µm orifice optimized for consistent performance in research settings. Its unique design ensures precise positioning, enhancing sorting accuracy and reliability. The product is ideal for users seeking high-performance solutions in cell analysis and sorting, although it is now discontinued.