Learn More
Cole-Parmer Essentials Aluminum Crimped-Walled Weighing Dishes with Tab, 70 mm dia, 60 mL; 100/PK
Catalog No. NC1969919 Shop All Cole-Parmer Products
Encompass_Preferred
Crimped sides for rigidity, sturdy gauge for heavy duty use. Large tab for sample numbers, flat bottom to improve sample handling, sides are angled for easy seperation. No oil residue, no vinyl coating to create contamination problems. Smooth bottom and may be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical.
Catalog No. NC1969919 Supplier Cole-Parmer Supplier No. 0101852
Crimped sides for rigidity, sturdy gauge for heavy duty use. Large tab for sample numbers, flat bottom to improve sample handling, sides are angled for easy seperation. No oil residue, no vinyl coating to create contamination problems. Smooth bottom and may be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical.