Learn More
Savillex Corporation Savillex 70 mm Closure, PFA Material, Leakproof Seal, 70 mm
SDP
Catalog No. NC0131263
Encompass
The Savillex Corporation 70 mm Closure is designed for demanding laboratory and industrial applications. Engineered from PFA, this closure provides exceptional chemical resistance and broad temperature compatibility, supporting temperatures from -328°F to 500°F. Its leakproof seal and external grips facilitate easy handling, ensuring secure sealing for various containers. This closure stands out in its class due to its robust construction and user-friendly design, making it the ideal choice for professionals seeking reliable performance.
Catalog No. NC0131263 Supplier Savillex Corporation Supplier No. 60007001
The Savillex Corporation 70 mm Closure is designed for demanding laboratory and industrial applications. Engineered from PFA, this closure provides exceptional chemical resistance and broad temperature compatibility, supporting temperatures from -328°F to 500°F. Its leakproof seal and external grips facilitate easy handling, ensuring secure sealing for various containers. This closure stands out in its class due to its robust construction and user-friendly design, making it the ideal choice for professionals seeking reliable performance.