Learn More
Newcomer Supply Alcohol, Reagent ACS, 70%, 1 gallon
SDP
Catalog No. NC2067204
Encompass
Shelf Life is 3 years from date of manufacture. Store at 15-30°C.
Catalog No. NC2067204 Supplier Newcomer Supply Supplier No. 10054C
Shelf Life is 3 years from date of manufacture. Store at 15-30°C.