Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

70% Reagent Alcohol in Conical Slide Holders, Cenmed™
SDP

Catalog No. 21520249
Change view
Click to view available options
Quantity:
100 x 25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-249 100 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-249 Supplier Cen-Med Enterprises Supplier No. 21520249
 Restricted Item
Add to Cart
Add to Cart

Enhance precision in your lab work with 70% Reagent Alcohol in Conical Slide Holders, specifically crafted for meticulous laboratory applications to ensure accurate results.

  • Unique formulation of 70% reagent alcohol maximizes efficiency in lab processes, making it an essential solution for tasks requiring a high level of accuracy
  • Each package includes conical slide holders, facilitating seamless handling and organization of your samples for a more efficient workflow
  • Combines the purity of reagent alcohol with the convenience of pre-packaged 25 mL conical slide holders, saving valuable time and effort
  • Save time and increase productivity with the all-in-one solution that simplifies your laboratory practices, from sample preparation to experimentation
  • Ensure precision and consistency in every experiment with a product crafted to meet the demands of critical laboratory work
  • Rely on quality and efficiency to meet the rigorous standards of your essential laboratory applications

Specifications

Physical Form Aqueous Solution, Liquid
CAS 67-63-0
Packaging Conical Slide Holder
Color Clear
Quantity 100 x 25 mL
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.