Learn More
MSC 3X110YD CLR #700 HOT MELT TAPE
Catalog No. 501919322
Encompass
3" x 110 Yd Clear Hot Melt Adhesive Packaging Tape Polypropylene Film Backing, 1.9 mil Thick, 22 Lb Tensile Strength
Catalog No. 50-191-9322 Supplier MSC Supplier No. 39589353
3" x 110 Yd Clear Hot Melt Adhesive Packaging Tape Polypropylene Film Backing, 1.9 mil Thick, 22 Lb Tensile Strength