Learn More
ABclonal Technology 700 Rabbit anti-Human IgD
Supplier: ABclonal Technology A26482500T
IgD is the major antigen receptor isotype on the surface of most peripheral B-cells where it is coexpressed with IgM The membrane-bound IgD (mIgD) induces the phosphorylation of CD79A and CD79B by the Src family of protein tyrosine kinases