Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ 7000 Series Pre-Assembled Multi-Gas Vapor Smart™ Cartridge Respirators
SDP

Catalog No. 19384329 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-384-329 Small
19-384-330 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-384-329 Supplier Moldex™ Supplier No. 7601
Add to Cart
Add to Cart

Pre-assembled respirator kits simplify inventory and are ideal for use against multiple hazards in the same workplace. Includes 7000 Series facepiece and 7600 Multi-Gas/Vapor Smart Cartridges.

  • Pre-assembled kit includes 7000 Series facepiece and 7600 Multi-Gas/Vapor Smart cartridges
  • Use with No. 8910 N95, 8970 R95, or 7940 P100 filters against particulates
  • Simplifies inventory by reducing the need to stock multiple cartridge types
  • Approved for 9 common gas/vapors
  • Cartridge selection made simpler and misuse reduced
  • Replaces up to five gas/vapor cartridges
  • 7000 facepiece features sleek design for modern, high-tech look and increased worker acceptance
  • Lightweight with extra-wide sealing area provides all-day comfort
  • Low profile, compact design fits well under welding helmets and with safety glasses
  • Mask drops down for convenient storage around the neck or locks down for custom fit
  • Adjustable head cradle and curved neck buckles for extra comfort
  • Wide field of vision
  • Maintenance made simple with very few replacement parts

Specifications

Cartridge Type Multi-Gas, Vapor Smart™ Cartridges
Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Material Thermoplastic Elastomer
Product Type Pre-Assembled Half-Mask Respirator
Size Small
Facepiece/Headtop Model 7000 series facepiece
For Use With (Application) Degreasing, forging, foundries, heat treating operations, paper making, rendering, rubber manufacturing
Includes 7000 Facepiece, 7600 Multi-Gas, Vapor Smart™ Cartridges
Model 7601
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.