Learn More
Sino Biological Recombinant Canine TIM-1/HAVCR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 70001D07H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of canine HAVCR1 (XP_854888.1) (Tyr 21-Ser 160) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3291