Learn More
Sino Biological Recombinant Canine IL-33 Protein
Supplier:  Sino Biological 70005DNAE20
Encompass_Preferred
A DNA sequence encoding the mature form of canine IL33 (O97863) (Ser 110-Ser 263) was expressed, with an initial Met at the N-terminus.
Catalog No. 50-162-3299