Learn More
Sino Biological Recombinant Canine IL-8/CXCL8 Protein
Supplier:  Sino Biological 70009DNAE50
Encompass_Preferred
A DNA sequence encoding the canine IL8 (P41324) (Ala23-Pro101) was expressed, with a N-terminal Met.
Catalog No. 50-162-3308