Learn More
Electron Microscopy Sciences EMS Vibrating Microtome Programable
SDP
Catalog No. 50949937
Encompass_Preferred
The 7000smz-2 unit has a submicron Z-axis deflection, a blade advance that is controllable to 10 microns/second, and a section thickness step size of 0.001 mm. It delivers perfect sections every time across a wide range of vibration speeds and amplitudes. Sub-Micron z-axis deflection and a blade advance controllable to 10 microns/second. All types of sectioning is possible.
Catalog No. 50-949-937 Supplier Electron Microscopy Sciences Supplier No. 7000SMZ2
Add to Cart
Add to Cart
Request a Quote
The 7000smz-2 unit has a submicron Z-axis deflection, a blade advance that is controllable to 10 microns/second, and a section thickness step size of 0.001 mm. It delivers perfect sections every time across a wide range of vibration speeds and amplitudes. Sub-Micron z-axis deflection and a blade advance controllable to 10 microns/second. All types of sectioning is possible.