Learn More
Sino Biological Recombinant Canine HE4/WFDC2 Protein (hFc Tag)
Supplier:  Sino Biological 70012D02H100
Encompass_Preferred
A DNA sequence encoding the canine WFDC2 (NP_001003241.1)(Met1-Phe124) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3309