Learn More
Sino Biological Recombinant Canine EGF Protein (His Tag)
Supplier:  Sino Biological 70013D07E1
Encompass_Preferred
A DNA sequence encoding the mature form of canine EGF (Q9BEA0) (Asn 973-Arg 1024) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3311