Learn More
Sino Biological Recombinant Canine EGF Protein
Supplier:  Sino Biological 70013DNAY1
Encompass_Preferred
A DNA sequence encoding the canine EGF (NP_001003094.1) (Asn973-Arg1024) was expressed.
Catalog No. 50-162-3314