Learn More
Sino Biological Recombinant Canine IL-2 Protein (147 Cys/Ser)
Supplier:  Sino Biological 70014DNAE50
Encompass_Preferred
A DNA sequence encoding the mature form of canine IL2 (Q29416) (Ala 21-Thr 155, 147Cys/Ser) was expressed, with an initial Met at the N-terminus.
Catalog No. 50-162-3318